Assay Details
| Gene Symbol: ACHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000665.4 | NP_000656.1 | 4 - 5 | 1915 | 105 | |
| NM_001282449.1 | NP_001269378.1 | 4 - 5 | 1651 | 105 | ||
| NM_001302622.1 | NP_001289551.1 | 4 - 5 | 1820 | 105 | ||
| XM_017012219.1 | XP_016867708.1 | 3 - 4 | 2271 | 105 | ||
| XM_017012220.1 | XP_016867709.1 | 3 - 4 | 2007 | 105 | ||
| GenBank mRNA | AF334270.1 | - | 4 - 5 | 1566 | 105 | |
| AK223443.1 | - | 4 - 5 | 1829 | 105 | ||
| AK291321.1 | - | 4 - 5 | 1815 | 105 | ||
| BC026315.1 | - | 4 - 5 | 1832 | 105 | ||
| M55040.1 | - | 3 - 4 | 1881 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001680.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
acetylcholine catabolic process in synaptic cleft
regulation of receptor recycling
osteoblast development
DNA replication
acetylcholine catabolic process
phosphatidylcholine biosynthetic process
cell adhesion
nervous system development
synapse assembly
muscle organ development
cell proliferation
response to wounding
receptor internalization
negative regulation of synaptic transmission, cholinergic
neurotransmitter biosynthetic process
amyloid precursor protein metabolic process
neurotransmitter receptor biosynthetic process
positive regulation of protein secretion
protein tetramerization
retina development in camera-type eye
beta-amyloid binding
acetylcholinesterase activity
cholinesterase activity
protein binding
collagen binding
hydrolase activity
serine hydrolase activity
acetylcholine binding
protein homodimerization activity
laminin binding
protein self-association
acetylcholine catabolic process in synaptic cleft
regulation of receptor recycling
osteoblast development
DNA replication
acetylcholine catabolic process
phosphatidylcholine biosynthetic process
cell adhesion
nervous system development
synapse assembly
muscle organ development
cell proliferation
response to wounding
receptor internalization
negative regulation of synaptic transmission, cholinergic
neurotransmitter biosynthetic process
amyloid precursor protein metabolic process
neurotransmitter receptor biosynthetic process
positive regulation of protein secretion
protein tetramerization
retina development in camera-type eye
beta-amyloid binding
acetylcholinesterase activity
cholinesterase activity
protein binding
collagen binding
hydrolase activity
serine hydrolase activity
acetylcholine binding
protein homodimerization activity
laminin binding
protein self-association
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000241069.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000302913.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000411582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000412389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000419336.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000426415.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000426415.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000426415.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000426415.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000428317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000430554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000430554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000430554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000430554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000430554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000440755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000440755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000440755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000440755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000440755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000442452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000442452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000442452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000442452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000442452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000442452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000454485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000454485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000454485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000454485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000454485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | ENST00000454485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_000665.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001282449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_001302622.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013599.hg.1 | NM_015831.2 | Refseq |

ACHE
Genomic Map