Assay Details
| Gene Symbol: ANKMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020319.2 | NP_064715.1 | 7 - 8 | 1127 | 65 | |
| XM_011515458.1 | XP_011513760.1 | 8 - 9 | 1254 | 65 | ||
| GenBank mRNA | AK001389.1 | - | 8 - 9 | 1096 | 65 | |
| AK001740.1 | - | 7 - 8 | 1065 | 65 | ||
| AK023704.1 | - | 7 - 8 | 1064 | 65 | ||
| AL050390.1 | - | 7 - 8 | 1069 | 65 | ||
| BC015453.1 | - | 1 - 2 | 45 | 65 | ||
| BC035353.1 | - | 7 - 8 | 1083 | 5185501 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001161.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018169.hg.1 | TC07001161.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018169.hg.1 | ENST00000306999.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018169.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173577.hg.1 | TC0700010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |

ANKMY2
Genomic Map