Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001744.hg.1 |
TC09000170.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001744.hg.1 |
ENST00000337747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001744.hg.1 |
ENST00000379158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001744.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
TC09000170.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
ENST00000337747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
ENST00000346365.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
ENST00000379154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
ENST00000379155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
ENST00000379158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001749.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147617.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147617.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147617.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147617.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147617.hg.1 |
TC0900007038.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147622.hg.1 |
TC0900007038.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000346365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000379158.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
ENST00000618590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_001244390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007038.hg.1 |
NM_147173.2 |
Refseq |