Assay Details
| Gene Symbol: CCNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017002421.1 | XP_016857910.1 | 6 - 7 | 797 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017002421.1 transcripts.
Set Membership: |
|

CCNL2
Genomic Map