Assay Details
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152540.3 | NP_689753.2 | 1 - 2 | 967 | 149 | |
XM_011534375.2 | XP_011532677.1 | 1 - 2 | 1003 | 149 | ||
XM_011534376.2 | XP_011532678.1 | 1 - 2 | 1003 | 149 | ||
XM_011534378.2 | XP_011532680.1 | 1 - 2 | 1003 | 149 | ||
XM_017007786.1 | XP_016863275.1 | 1 - 2 | 1003 | 149 | ||
XM_017007787.1 | XP_016863276.1 | 1 - 2 | 1003 | 149 | ||
GenBank mRNA | AB074276.1 | - | 1 - 2 | 940 | 149 | |
AK096833.1 | - | 1 - 2 | 953 | 149 | ||
AY299407.1 | - | 1 - 2 | 967 | 149 | ||
BC012458.1 | - | 1 - 2 | 733 | 149 | ||
BC021137.1 | - | 1 - 2 | 924 | 4866354 | 149 | |
BC032453.1 | - | 1 - 2 | 965 | 5176029 | 149 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001197.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04016422.hg.1 | TC04001197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016422.hg.1 | ENST00000388940.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016422.hg.1 | ENST00000401642.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016422.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04016425.hg.1 | TC04001197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016425.hg.1 | ENST00000388940.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016425.hg.1 | ENST00000401642.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016425.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04016424.hg.1 | TC04001197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016424.hg.1 | ENST00000388940.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016424.hg.1 | ENST00000401642.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016424.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04016421.hg.1 | TC04001197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016421.hg.1 | ENST00000388940.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016421.hg.1 | ENST00000401642.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016421.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04016423.hg.1 | TC04001197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016423.hg.1 | ENST00000388940.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016423.hg.1 | ENST00000401642.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016423.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172864.hg.1 | TC0400010699.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172860.hg.1 | TC0400010699.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172862.hg.1 | TC0400010699.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172863.hg.1 | TC0400010699.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172861.hg.1 | TC0400010699.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172865.hg.1 | TC0400010699.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000388940.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000401642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000503450.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | ENST00000611795.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010699.hg.1 | NM_152540.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 137317 | - | - |
