Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016420.hg.1 |
TC04001197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016420.hg.1 |
ENST00000388940.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016420.hg.1 |
ENST00000401642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016420.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172859.hg.1 |
TC0400010699.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000388940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000401642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000503450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
ENST00000611795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010699.hg.1 |
NM_152540.3 |
Refseq |