Assay Details
Gene Symbol: KCNH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139318.4 | NP_647479.2 | 7 - 8 | 1423 | 99 | |
NM_172375.2 | NP_758963.1 | 7 - 8 | 1423 | 99 | ||
GenBank mRNA | AF418206.1 | - | 7 - 8 | 1371 | 99 | |
AF472412.1 | - | 7 - 8 | 1371 | 99 | ||
AF493798.1 | - | 7 - 8 | 1371 | 99 | ||
BC073979.1 | - | 7 - 8 | 1423 | 6199697 | 99 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001202.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
regulation of G2/M transition of mitotic cell cycle
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
phosphorelay sensor kinase activity
voltage-gated potassium channel activity
calmodulin binding
ion channel binding
protein heterodimerization activity
phosphorelay signal transduction system
regulation of G2/M transition of mitotic cell cycle
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
phosphorelay sensor kinase activity
voltage-gated potassium channel activity
calmodulin binding
ion channel binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14013716.hg.1 | ENST00000322893.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013716.hg.1 | ENST00000394968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013716.hg.1 | ENST00000420622.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013716.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14013716.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14013716.hg.1 | TC14001202.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | ENST00000322893.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | ENST00000394968.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | ENST00000420622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | NM_139318.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | NM_172375.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169303.hg.1 | TC1400009381.hg.1 | Clariom D TC |
