Assay Details
Gene Symbol: IFI27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130080.2 | NP_001123552.1 | 1 - 2 | 63 | 71 | |
NM_005532.4 | NP_005523.3 | 1 - 2 | 63 | 71 | ||
GenBank mRNA | AK289535.1 | - | 1 - 2 | 61 | 71 | |
BM841596.1 | - | 1 - 2 | 60 | 71 | ||
BU601451.1 | - | 1 - 2 | 63 | 71 | ||
BU688913.1 | - | 4 - 5 | 619 | 71 | ||
BX647330.1 | - | 1 - 2 | 60 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000584.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK289535.1, NM_001130080.2, NM_005532.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
activation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein export from nucleus
type I interferon signaling pathway
apoptotic signaling pathway
RNA polymerase II activating transcription factor binding
molecular_function
lamin binding
negative regulation of transcription from RNA polymerase II promoter
activation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein export from nucleus
type I interferon signaling pathway
apoptotic signaling pathway
RNA polymerase II activating transcription factor binding
molecular_function
lamin binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000611954.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000611954.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000611954.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 152567 | - | - |
