Assay Details
| Gene Symbol: CAPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004058.4 | NP_004049.2 | 2 - 3 | 769 | 55 | |
| NM_080590.3 | NP_542157.2 | 2 - 3 | 769 | 55 | ||
| GenBank mRNA | AK090469.1 | - | 2 - 3 | 3053 | 55 | |
| AK226026.1 | - | 231 | 55 | |||
| BC019065.1 | - | 182 | 55 | |||
| BC080599.1 | - | 2 - 3 | 177 | 4876766 | 55 | |
| BC101513.1 | - | 1 - 2 | 106 | 8069019 | 55 | |
| BC104766.1 | - | 1 - 2 | 106 | 8143769 | 55 | |
| BG768134.1 | - | 2 - 3 | 206 | 55 | ||
| X97966.1 | - | 1 - 2 | 81 | 55 | ||
| Gene Symbol: VMAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DA541168.1 | - | 3 - 4 | 437 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | TC1900011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900198951.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000222125.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000452990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000585541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000588865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000590428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | ENST00000618299.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_004058.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011648.hg.1 | NM_080590.3 | Refseq |

CAPS
Genomic Map