| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02014685.hg.1 |
TC02005000.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02014685.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02014685.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02014685.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200224011.hg.1 |
TC0200016560.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000284669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000463400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
ENST00000480330.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016560.hg.1 |
NM_006063.2 |
Refseq |