Assay Details
| Gene Symbol: FGF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033163.3 | NP_149353.1 | 3 - 4 | 256 | 105 | |
| GenBank mRNA | AB014615.1 | - | 3 - 4 | 163 | 105 | |
More Information
Set Membership: |
|

FGF8
Genomic Map