Assay Details
Gene Symbol: EXOSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318363.1 | NP_001305292.1 | 3 - 4 | 253 | 88 | |
NM_001318365.1 | NP_001305294.1 | 4 - 5 | 407 | 88 | ||
NM_001318366.1 | NP_001305295.1 | 3 - 4 | 253 | 88 | ||
NM_016046.4 | NP_057130.1 | 3 - 4 | 253 | 88 | ||
NR_134586.1 | 3 - 4 | 253 | 88 | |||
XM_011539847.2 | XP_011538149.1 | 2 - 3 | 228 | 88 | ||
GenBank mRNA | AF151866.1 | - | 3 - 4 | 238 | 88 | |
AK313717.1 | - | 3 - 4 | 241 | 88 | ||
BC022067.2 | - | 3 - 4 | 228 | 4817073 | 88 | |
BG331954.1 | - | 4 - 5 | 366 | 88 | ||
BI550688.1 | - | 3 - 4 | 258 | 88 | ||
BM692452.1 | - | 3 - 4 | 228 | 88 | ||
BU569997.1 | - | 3 - 4 | 245 | 88 | ||
DB137484.1 | - | 3 - 4 | 247 | 88 | ||
N34159.1 | - | 3 - 4 | 207 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001566.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001318365.1,NM_001318366.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000474309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000474309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000474309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10020005.hg.1 | ENST00000370885.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10020005.hg.1 | ENST00000370886.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10020005.hg.1 | ENST00000370902.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10020005.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10020005.hg.1 | TC10001566.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370884.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000474309.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000474309.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000474309.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | ENST00000489158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011552.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000370885.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000370886.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000370902.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000471049.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000472345.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000476234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000477692.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | ENST00000485122.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | NM_016046.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000178621.hg.1 | TC1000011552.hg.1 | Clariom D TC |
