Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023572.hg.1 |
TC04001705.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023572.hg.1 |
ENST00000306480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023572.hg.1 |
ENST00000506087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023572.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191305.hg.1 |
TC0400012989.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000505095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
ENST00000506087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012989.hg.1 |
NM_001100389.1 |
Refseq |