Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000280772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000373817.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000373827.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000395293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000395299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000395304.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000460468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000503366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
ENST00000536348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
NM_001204403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
NM_001204404.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
NM_020987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016621.hg.1 |
TC10001319.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000280772.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000373827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000460468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000474360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000503366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000622427.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
NM_001204403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
NM_001204404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
NM_020987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000280772.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000373827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000460468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000474360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000503366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
ENST00000622427.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
NM_001204403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
NM_001204404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
NM_020987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173220.hg.1 |
TC1000010727.hg.1 |
Clariom D TC |