Assay Details
| Gene Symbol: RAB7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164522.2 | NP_001157994.1 | 3 - 4 | 410 | 107 | |
| NM_001304839.1 | NP_001291768.1 | 3 - 4 | 410 | 107 | ||
| NM_177403.5 | NP_796377.3 | 2 - 3 | 484 | 107 | ||
| XM_006711288.3 | XP_006711351.1 | 3 - 4 | 407 | 107 | ||
| GenBank mRNA | AK122657.1 | - | 3 - 4 | 405 | 107 | |
| AK127140.1 | - | 2 - 3 | 468 | 107 | ||
| AY094596.1 | - | 3 - 4 | 357 | 107 | ||
| BC017092.1 | - | 3 - 4 | 352 | 3851998 | 107 | |
| BI757251.1 | - | 2 - 3 | 481 | 107 | ||
| DA966224.1 | - | 3 - 4 | 410 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
protein transport
positive regulation of interleukin-6 production
negative regulation of toll-like receptor 4 signaling pathway
negative regulation of toll-like receptor 9 signaling pathway
late endosome to Golgi transport
positive regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
GTP binding
small GTPase mediated signal transduction
protein transport
positive regulation of interleukin-6 production
negative regulation of toll-like receptor 4 signaling pathway
negative regulation of toll-like receptor 9 signaling pathway
late endosome to Golgi transport
positive regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
GTP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229517.hg.1 | TC0100017083.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |

RAB7B
Genomic Map