Assay Details
Gene Symbol: PAWR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002583.2 | NP_002574.2 | 4 - 5 | 972 | 136 | |
XM_006719435.2 | XP_006719498.1 | 4 - 5 | 948 | 136 | ||
XM_017019377.1 | XP_016874866.1 | 4 - 5 | 852 | 136 | ||
GenBank mRNA | BC007018.1 | - | 3 - 4 | 818 | 3828412 | 136 |
CR536549.1 | - | 3 - 4 | 686 | 136 | ||
U63809.1 | - | 4 - 5 | 930 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001765.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
apoptotic process
response to wounding
response to iron(II) ion
negative regulation of B cell proliferation
response to lipopolysaccharide
positive regulation of phosphoprotein phosphatase activity
interleukin-2 biosynthetic process
negative regulation of T cell proliferation
positive regulation of amyloid precursor protein biosynthetic process
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
negative regulation of T cell receptor signaling pathway
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
actin filament bundle assembly
positive regulation of hindgut contraction
cellular response to vitamin E
cellular response to interleukin-1
cellular response to follicle-stimulating hormone stimulus
cellular response to estradiol stimulus
negative regulation of calcium ion import
apoptotic signaling pathway
activation of cysteine-type endopeptidase activity
positive regulation of relaxation of smooth muscle
positive regulation of leukocyte tethering or rolling
positive regulation of neuronal action potential
calcium ion import into cell
positive regulation of neutrophil extravasation
transcription corepressor activity
actin binding
protein kinase C binding
protein binding
protein phosphatase 1 binding
enzyme binding
leucine zipper domain binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
apoptotic process
response to wounding
response to iron(II) ion
negative regulation of B cell proliferation
response to lipopolysaccharide
positive regulation of phosphoprotein phosphatase activity
interleukin-2 biosynthetic process
negative regulation of T cell proliferation
positive regulation of amyloid precursor protein biosynthetic process
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
negative regulation of T cell receptor signaling pathway
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
actin filament bundle assembly
positive regulation of hindgut contraction
cellular response to vitamin E
cellular response to interleukin-1
cellular response to follicle-stimulating hormone stimulus
cellular response to estradiol stimulus
negative regulation of calcium ion import
apoptotic signaling pathway
activation of cysteine-type endopeptidase activity
positive regulation of relaxation of smooth muscle
positive regulation of leukocyte tethering or rolling
positive regulation of neuronal action potential
calcium ion import into cell
positive regulation of neutrophil extravasation
transcription corepressor activity
actin binding
protein kinase C binding
protein binding
protein phosphatase 1 binding
enzyme binding
leucine zipper domain binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12023323.hg.1 | ENST00000328827.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12023323.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12023323.hg.1 | TC12001765.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200188021.hg.1 | TC1200011364.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000328827.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000547699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548075.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000548426.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000549050.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000550603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000551712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | ENST00000552637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011364.hg.1 | NM_002583.3 | Refseq |
