Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024887.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024887.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024887.hg.1 |
TC17001858.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011699.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194695.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194695.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194695.hg.1 |
ENST00000330793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194695.hg.1 |
NM_006678.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194695.hg.1 |
TC1700011699.hg.1 |
Clariom D TC |