Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007662.hg.1 |
TC03000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007662.hg.1 |
ENST00000330452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007662.hg.1 |
ENST00000394729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007662.hg.1 |
NM_006254.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007662.hg.1 |
NM_212539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
ENST00000330452.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
ENST00000394729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
NM_006254.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
NM_212539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
ENST00000330452.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
ENST00000394729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
NM_006254.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
NM_212539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154307.hg.1 |
TC0300007512.hg.1 |
Clariom D TC |