Assay Details
Gene Symbol: SMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005631.4 | NP_005622.1 | 3 - 4 | 1032 | 58 | |
XM_011516522.2 | XP_011514824.1 | 4 - 5 | 598 | 58 | ||
GenBank mRNA | AF071494.1 | - | 3 - 4 | 752 | 58 | |
AF120103.1 | - | 3 - 4 | 810 | 58 | ||
BC009989.2 | - | 3 - 4 | 1032 | 4127774 | 58 | |
U84401.1 | - | 3 - 4 | 752 | 58 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000795.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
osteoblast differentiation
in utero embryonic development
cell fate specification
neural crest cell migration
heart looping
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
determination of left/right asymmetry in lateral mesoderm
type B pancreatic cell development
G-protein coupled receptor signaling pathway
smoothened signaling pathway
positive regulation of hh target transcription factor activity
ventral midline determination
midgut development
anterior/posterior pattern specification
positive regulation of gene expression
negative regulation of gene expression
dentate gyrus development
cerebellar cortex morphogenesis
thalamus development
dorsal/ventral neural tube patterning
smoothened signaling pathway involved in ventral spinal cord patterning
smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
central nervous system neuron differentiation
cerebral cortex development
negative regulation of epithelial cell differentiation
hair follicle morphogenesis
protein localization to nucleus
multicellular organism growth
non-canonical Wnt signaling pathway
positive regulation of multicellular organism growth
positive regulation of protein import into nucleus
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
negative regulation of DNA binding
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
positive regulation of organ growth
astrocyte activation
skeletal muscle fiber development
forebrain morphogenesis
homeostasis of number of cells within a tissue
positive regulation of epithelial cell proliferation
protein stabilization
myoblast migration
negative regulation of hair follicle development
canonical Wnt signaling pathway
detection of cell density by contact stimulus involved in contact inhibition
atrial septum morphogenesis
mammary gland epithelial cell differentiation
epithelial-mesenchymal cell signaling
somite development
pancreas morphogenesis
left/right axis specification
cellular response to cholesterol
renal system development
mesenchymal to epithelial transition involved in metanephric renal vesicle formation
positive regulation of branching involved in ureteric bud morphogenesis
regulation of stem cell population maintenance
regulation of heart morphogenesis
G-protein coupled receptor activity
patched binding
protein binding
drug binding
Wnt-protein binding
Wnt-activated receptor activity
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
osteoblast differentiation
in utero embryonic development
cell fate specification
neural crest cell migration
heart looping
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
determination of left/right asymmetry in lateral mesoderm
type B pancreatic cell development
G-protein coupled receptor signaling pathway
smoothened signaling pathway
positive regulation of hh target transcription factor activity
ventral midline determination
midgut development
anterior/posterior pattern specification
positive regulation of gene expression
negative regulation of gene expression
dentate gyrus development
cerebellar cortex morphogenesis
thalamus development
dorsal/ventral neural tube patterning
smoothened signaling pathway involved in ventral spinal cord patterning
smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
central nervous system neuron differentiation
cerebral cortex development
negative regulation of epithelial cell differentiation
hair follicle morphogenesis
protein localization to nucleus
multicellular organism growth
non-canonical Wnt signaling pathway
positive regulation of multicellular organism growth
positive regulation of protein import into nucleus
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
negative regulation of DNA binding
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
positive regulation of organ growth
astrocyte activation
skeletal muscle fiber development
forebrain morphogenesis
homeostasis of number of cells within a tissue
positive regulation of epithelial cell proliferation
protein stabilization
myoblast migration
negative regulation of hair follicle development
canonical Wnt signaling pathway
detection of cell density by contact stimulus involved in contact inhibition
atrial septum morphogenesis
mammary gland epithelial cell differentiation
epithelial-mesenchymal cell signaling
somite development
pancreas morphogenesis
left/right axis specification
cellular response to cholesterol
renal system development
mesenchymal to epithelial transition involved in metanephric renal vesicle formation
positive regulation of branching involved in ureteric bud morphogenesis
regulation of stem cell population maintenance
regulation of heart morphogenesis
G-protein coupled receptor activity
patched binding
protein binding
drug binding
Wnt-protein binding
Wnt-activated receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07012528.hg.1 | TC07000795.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07012528.hg.1 | ENST00000249373.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07012528.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700164636.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700164636.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700164636.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700164636.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700164636.hg.1 | TC0700009082.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000249373.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000462420.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000475779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | ENST00000495998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009082.hg.1 | NM_005631.4 | Refseq |
