Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012535.hg.1 |
TC07000795.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012535.hg.1 |
ENST00000249373.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012535.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164644.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164644.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164644.hg.1 |
ENST00000249373.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164644.hg.1 |
NM_005631.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164644.hg.1 |
TC0700009082.hg.1 |
Clariom D TC |