Assay Details
| Gene Symbol: RBBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006722519.2 | XP_006722582.1 | 3 - 4 | 298 | 143 | |
| XM_006722520.2 | XP_006722583.1 | 2 - 3 | 234 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006722519.2, XM_006722520.2 transcripts.
Set Membership: |
|

RBBP8
Genomic Map