Assay Details
Gene Symbol: GMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003875.2 | NP_003866.1 | 1 - 2 | 364 | 63 | |
GenBank mRNA | AK223399.1 | - | 1 - 2 | 247 | 63 | |
AK291504.1 | - | 1 - 2 | 232 | 63 | ||
AK300787.1 | - | 1 - 2 | 247 | 63 | ||
AK315832.1 | - | 2 - 3 | 139 | 63 | ||
BC012178.1 | - | 1 - 2 | 232 | 4653684 | 63 | |
BG721087.1 | - | 1 - 2 | 369 | 63 | ||
U10860.1 | - | 1 - 2 | 151 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000851.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300787.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03015017.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015017.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015017.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015025.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015025.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015025.hg.1 | ENST00000541628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015025.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015021.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015021.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015021.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015021.hg.1 | ENST00000537975.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015021.hg.1 | ENST00000541628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015021.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015023.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015023.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015023.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015023.hg.1 | ENST00000537975.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015023.hg.1 | ENST00000541628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015023.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015022.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015022.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015022.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015022.hg.1 | ENST00000537975.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015022.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015020.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015020.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015020.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015020.hg.1 | ENST00000537975.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015020.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015024.hg.1 | TC03000851.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015024.hg.1 | ENST00000496455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015024.hg.1 | ENST00000537975.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015024.hg.1 | ENST00000541628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015024.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166411.hg.1 | TC0300009271.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166412.hg.1 | TC0300009271.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000295920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000476145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009271.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166415.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166415.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166415.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166415.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166415.hg.1 | TC0300009271.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300166409.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166409.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166409.hg.1 | ENST00000496455.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166409.hg.1 | NM_003875.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166409.hg.1 | TC0300009271.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 115522 | - | - |
