| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034355.hg.1 |
TC03001906.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034355.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034355.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034356.hg.1 |
TC03001906.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034356.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034356.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034357.hg.1 |
TC03001906.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034357.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034357.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012815.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012815.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012815.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012815.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012815.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012815.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012815.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012815.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012815.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012815.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012815.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012815.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195918.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195918.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195918.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195918.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195918.hg.1 |
TC0300012815.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300195919.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195919.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195919.hg.1 |
ENST00000325602.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300195919.hg.1 |
NM_176894.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300195919.hg.1 |
TC0300012815.hg.1 |
Clariom D TC |