Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025646.hg.1 |
TC06004055.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025646.hg.1 |
ENST00000368507.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025646.hg.1 |
ENST00000368508.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025646.hg.1 |
NM_002944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
ENST00000368507.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
ENST00000368508.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
NM_002944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
ENST00000368507.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
ENST00000368508.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
NM_002944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202551.hg.1 |
TC0600014336.hg.1 |
Clariom D TC |