Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007145.2 | NP_009076.2 | 1 - 2 | 590 | 97 | |
GenBank mRNA | BC065568.1 | - | 1 - 2 | 59 | 97 | |
DA395000.1 | - | 1 - 2 | 86 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000497.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_007145.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19006625.hg.1 | ENST00000443387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19006625.hg.1 | ENST00000456324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19006625.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19006625.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19006625.hg.1 | TC19000497.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19006626.hg.1 | ENST00000443387.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19006626.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19006626.hg.1 | TC19000497.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | TC1900007917.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159055.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000456324.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000586094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000587285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | ENST00000591063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099638.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_001099639.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007917.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159056.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159056.hg.1 | NM_007145.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159056.hg.1 | TC1900007917.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900159056.hg.1 | ENST00000443387.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159056.hg.1 | NM_007145.2 | Refseq |
