Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020642.hg.1 |
TC02001326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020642.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020642.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177597.hg.1 |
TC0200010847.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000373960.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000477226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000483395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
ENST00000492726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010847.hg.1 |
NM_001927.3 |
Refseq |