Assay Details
| Gene Symbol: SLC9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF073299.1 | - | 9 - 10 | 1931 | 101 | |
| BC136377.1 | - | 9 - 10 | 2001 | 9020364 | 101 | |
| BC136378.1 | - | 9 - 10 | 2001 | 9020365 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160702.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160702.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160702.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160702.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160702.hg.1 | TC0200008680.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |

SLC9A2
Genomic Map