Assay Details
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020866.2 | NP_065917.1 | 1 - 2 | 1296 | 85 | |
GenBank mRNA | AB040923.1 | - | 1 - 2 | 1262 | 85 | |
AF252283.1 | - | 1 - 2 | 1130 | 85 | ||
AK054609.1 | - | 1 - 2 | 508 | 85 | ||
AK291435.1 | - | 1 - 2 | 1296 | 85 | ||
BC022460.1 | - | 1 - 2 | 540 | 4819077 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000721.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13008323.hg.1 | ENST00000377844.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13008323.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008323.hg.1 | TC13000721.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13008327.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008327.hg.1 | TC13000721.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008324.hg.1 | ENST00000377844.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13008324.hg.1 | ENST00000545028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13008324.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008324.hg.1 | TC13000721.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13008326.hg.1 | ENST00000377844.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13008326.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008326.hg.1 | TC13000721.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008325.hg.1 | ENST00000377844.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13008325.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13008325.hg.1 | TC13000721.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161930.hg.1 | TC1300009204.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161932.hg.1 | TC1300009204.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009204.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161929.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161929.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161929.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161929.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161929.hg.1 | TC1300009204.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | ENST00000377844.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161931.hg.1 | TC1300009204.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | ENST00000545028.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | NM_001286725.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | NM_020866.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300161933.hg.1 | TC1300009204.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000414504.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | ENST00000424524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007446.hg.1 | NR_002717.2 | Refseq |
