Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008320.hg.1 |
ENST00000377844.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008320.hg.1 |
ENST00000545028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008320.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008320.hg.1 |
TC13000721.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161926.hg.1 |
TC1300009204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000377844.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
ENST00000545028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_001286725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009204.hg.1 |
NM_020866.2 |
Refseq |