Assay Details
Gene Symbol: SNRPD2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033826.1 | 1 | 465 | 80 | ||
GenBank mRNA | BC014600.1 | - | 1 | 471 | 4826560 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | egl-9 family hypoxia inducible factor 1 | EGLN1 |
C1orf12, ECYT3, HALAH, HIF-PH2, HIFPH2, HPH-2, HPH2, PHD2, SM20, ZMYND6 |
-50.7kb | Sense | Chr. 1 - 231363751 - 231425044 on NCBI Build GRCh38 |
Downstream | translin associated factor X | TSNAX | TRAX | 52.1kb | Anti-sense | Chr. 1 - 231528653 - 231566524 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003969.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929609 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858990 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859026 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929592 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929590 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858963 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929595 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859019 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929583 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929632 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929619 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858956 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929604 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858981 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929620 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929613 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929586 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929596 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929617 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929593 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929579 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929624 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929581 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858939 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929634 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858949 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929569 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929610 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929627 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929625 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929618 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00859028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858991 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858936 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929555 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00858994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929567 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20929571 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42653 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3384900 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P494511 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7925059 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7925060 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7925061 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7925058 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01060101.hg.1 | TC01003969.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01060101.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017731.hg.1 | ENST00000473818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017731.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017731.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017731.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017731.hg.1 | ENST00000473818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017731.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017731.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017731.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233775.hg.1 | ENST00000473818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100233775.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233775.hg.1 | ENST00000473818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100233775.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233775.hg.1 | TC0100017731.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100233774.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233774.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233774.hg.1 | TC0100017731.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100233776.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233776.hg.1 | NR_033826.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100233776.hg.1 | TC0100017731.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012426 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012426 | - | - |
