Assay Details
| Gene Symbol: ASRGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083926.1 | NP_001077395.1 | 2 - 3 | 552 | 63 | |
| NM_025080.3 | NP_079356.3 | 2 - 3 | 475 | 63 | ||
| XM_011545265.2 | XP_011543567.1 | 2 - 3 | 434 | 63 | ||
| XM_011545266.2 | XP_011543568.1 | 2 - 3 | 434 | 63 | ||
| XM_017018354.1 | XP_016873843.1 | 2 - 3 | 434 | 63 | ||
| GenBank mRNA | AF411076.1 | - | 2 - 3 | 284 | 63 | |
| AK090908.1 | - | 2 - 3 | 324 | 63 | ||
| AK091894.1 | - | 2 - 3 | 406 | 63 | ||
| AK225802.1 | - | 2 - 3 | 243 | 63 | ||
| AK313069.1 | - | 2 - 3 | 335 | 63 | ||
| BC021295.2 | - | 2 - 3 | 273 | 3937243 | 63 | |
| BC093070.1 | - | 2 - 3 | 377 | 30370741 | 63 | |
| BP308677.1 | - | 2 - 3 | 475 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000551.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11006711.hg.1 | ENST00000301776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006711.hg.1 | ENST00000415229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006711.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006711.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006711.hg.1 | TC11000551.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155882.hg.1 | TC1100007844.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 195464 | - | - |

ASRGL1
Genomic Map