Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057625.hg.1 |
TC01003756.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057625.hg.1 |
ENST00000340781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057625.hg.1 |
ENST00000367134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057625.hg.1 |
ENST00000367135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057625.hg.1 |
NM_052934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057625.hg.1 |
NM_134325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000340781.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000367134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000367135.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000491127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
NM_052934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
NM_134325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000340781.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000367134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000367135.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
ENST00000491127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
NM_052934.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
NM_134325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229480.hg.1 |
TC0100017082.hg.1 |
Clariom D TC |