Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011948.hg.1 |
TC03000664.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011948.hg.1 |
ENST00000265056.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011948.hg.1 |
ENST00000468414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011948.hg.1 |
ENST00000539922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011948.hg.1 |
ENST00000543142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011948.hg.1 |
NM_004526.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000265056.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000468414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000474964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000477668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000491422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
NM_004526.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
NR_073375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000265056.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000468414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000474964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000477668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
ENST00000491422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
NM_004526.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
NR_073375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161817.hg.1 |
TC0300008702.hg.1 |
Clariom D TC |