Assay Details
| Gene Symbol: PPIH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540508.1 | XP_011538810.1 | 3 - 4 | 296 | 75 | |
| XM_017000058.1 | XP_016855547.1 | 3 - 4 | 950 | 75 | ||
| XM_017000059.1 | XP_016855548.1 | 3 - 4 | 296 | 75 | ||
| GenBank mRNA | AK294288.1 | - | 1 - 2 | 453 | 75 | |
| AK309438.1 | - | 3 - 4 | 292 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK294288.1, XM_011540508.1, XM_017000058.1, XM_017000059.1 transcripts.
Set Membership: |
|

PPIH
Genomic Map