Assay Details
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278716.1 | NP_001265645.1 | 1 - 2 | 200 | 74 | |
XM_005266930.2 | XP_005266987.1 | 1 - 2 | 188 | 74 | ||
XM_011535748.2 | XP_011534050.1 | 1 - 2 | 188 | 74 | ||
XM_017010726.1 | XP_016866215.1 | 1 - 2 | 132 | 74 | ||
XM_017010728.1 | XP_016866217.1 | 1 - 2 | 200 | 74 | ||
GenBank mRNA | BC091484.1 | - | 1 - 2 | 120 | 6178874 | 74 |
DA391493.1 | - | 1 - 2 | 188 | 74 | ||
DA782779.1 | - | 1 - 2 | 200 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091484.1,NM_001278716.1,XM_005266930.2,XM_011535748.2,XM_017010726.1,XM_017010728.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187120.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187120.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187120.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187120.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187120.hg.1 | TC0600012647.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187121.hg.1 | TC0600012647.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187123.hg.1 | TC0600012647.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 198970 | - | - |
