Assay Details
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278716.1 | NP_001265645.1 | 5 - 6 | 1360 | 117 | |
NM_012160.4 | NP_036292.2 | 4 - 5 | 1242 | 117 | ||
XM_005266930.2 | XP_005266987.1 | 5 - 6 | 1348 | 117 | ||
XM_011535748.2 | XP_011534050.1 | 5 - 6 | 1348 | 117 | ||
XM_017010726.1 | XP_016866215.1 | 4 - 5 | 1174 | 117 | ||
XM_017010727.1 | XP_016866216.1 | 4 - 5 | 1202 | 117 | ||
XM_017010728.1 | XP_016866217.1 | 6 - 7 | 1436 | 117 | ||
GenBank mRNA | AF174590.1 | - | 2 - 3 | 851 | 117 | |
AF176699.1 | - | 2 - 3 | 851 | 117 | ||
AF199355.1 | - | 2 - 3 | 855 | 117 | ||
AK313076.1 | - | 4 - 5 | 1171 | 117 | ||
BC055010.1 | - | 4 - 5 | 1181 | 6137889 | 117 | |
BC091484.1 | - | 5 - 6 | 1280 | 6178874 | 117 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001961.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06024153.hg.1 | TC06001961.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024153.hg.1 | ENST00000229971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024153.hg.1 | ENST00000369244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024153.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187114.hg.1 | TC0600012647.hg.1 | Clariom D TC |
