Assay Details
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278716.1 | NP_001265645.1 | 7 - 8 | 1827 | 88 | |
NM_012160.4 | NP_036292.2 | 6 - 7 | 1709 | 88 | ||
NR_103836.1 | 5 - 6 | 1363 | 88 | |||
XM_017010726.1 | XP_016866215.1 | 6 - 7 | 1641 | 88 | ||
XM_017010728.1 | XP_016866217.1 | 8 - 9 | 1903 | 88 | ||
GenBank mRNA | AF174590.1 | - | 4 - 5 | 1318 | 88 | |
AF176699.1 | - | 4 - 5 | 1318 | 88 | ||
AF199355.1 | - | 4 - 5 | 1322 | 88 | ||
AK313076.1 | - | 6 - 7 | 1638 | 88 | ||
BC055010.1 | - | 6 - 7 | 1648 | 6137889 | 88 | |
BC091484.1 | - | 7 - 8 | 1747 | 6178874 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001961.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06024150.hg.1 | TC06001961.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024150.hg.1 | ENST00000229971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024150.hg.1 | ENST00000369244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024150.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06024151.hg.1 | TC06001961.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06024151.hg.1 | ENST00000229971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024151.hg.1 | ENST00000369244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06024151.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600187110.hg.1 | TC0600012647.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000229971.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | ENST00000369244.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_001278716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NM_012160.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012647.hg.1 | NR_103837.1 | Refseq |
