Assay Details
| Gene Symbol: METTL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007239.1 | NP_001007240.1 | 2 - 3 | 796 | 73 | |
| GenBank mRNA | AL049669.1 | - | 2 - 3 | 721 | 73 | |
More Information
Set Membership: |
|

METTL13
Genomic Map