Assay Details
Gene Symbol: HAPLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001884.3 | NP_001875.1 | 1 - 2 | 281 | 94 | |
XM_017009053.1 | XP_016864542.1 | 1 - 2 | 264 | 94 | ||
GenBank mRNA | AK313713.1 | - | 1 - 2 | 236 | 94 | |
U43328.1 | - | 1 - 2 | 83 | 94 | ||
X17405.1 | - | 1 - 2 | 281 | 94 | ||
X78076.1 | - | 1 - 2 | 281 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001557.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313713.1,NM_001884.3,U43328.1,X17405.1,XM_017009053.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021252.hg.1 | TC05001557.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05021252.hg.1 | ENST00000274341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021252.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05021243.hg.1 | TC05001557.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05021243.hg.1 | ENST00000274341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021243.hg.1 | ENST00000514416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05021243.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178583.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178583.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178583.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178583.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178583.hg.1 | TC0500011347.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178572.hg.1 | TC0500011347.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000503117.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000504713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000508307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000510978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000514416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011347.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | ENST00000274341.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | ENST00000515590.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | NM_001884.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500178582.hg.1 | TC0500011347.hg.1 | Clariom D TC |
