Assay Details
| Gene Symbol: NPFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320296.1 | NP_001307225.1 | 1 - 2 | 617 | 117 | |
| NM_003717.3 | NP_003708.1 | 2 - 3 | 387 | 117 | ||
| GenBank mRNA | AF005271.1 | - | 2 - 3 | 386 | 117 | |
| BC104234.1 | - | 1 - 2 | 566 | 40033026 | 117 | |
| BC104235.1 | - | 1 - 2 | 566 | 40033028 | 117 | |
More Information
Set Membership: |
|

NPFF
Genomic Map