Assay Details
Gene Symbol: CLEC4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194450.2 | NP_919432.1 | 2 - 3 | 450 | 150 | |
GenBank mRNA | AF200738.1 | - | 2 - 3 | 203 | 150 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000119.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | ENST00000229332.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | ENST00000345999.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | ENST00000352620.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12001629.hg.1 | TC12000119.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199159.hg.1 | TC1200012593.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000229332.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000345999.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000352620.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000360500.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | ENST00000546339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_016184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194447.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194448.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012593.hg.1 | NM_194450.2 | Refseq |
