Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002808.hg.1 |
TC0X000238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002808.hg.1 |
ENST00000218388.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002808.hg.1 |
ENST00000377018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002808.hg.1 |
ENST00000456754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002808.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002811.hg.1 |
TC0X000238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002811.hg.1 |
ENST00000218388.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002811.hg.1 |
ENST00000377018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002811.hg.1 |
ENST00000456754.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002811.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002807.hg.1 |
TC0X000238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002807.hg.1 |
ENST00000218388.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002807.hg.1 |
ENST00000377018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002807.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148745.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148745.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148745.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148745.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148745.hg.1 |
TC0X00007149.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148746.hg.1 |
TC0X00007149.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00148744.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148744.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148744.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148744.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148744.hg.1 |
TC0X00007149.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148747.hg.1 |
TC0X00007149.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00148750.hg.1 |
TC0X00007149.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000218388.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000377017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000441738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000445623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
ENST00000456754.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007149.hg.1 |
NM_003254.2 |
Refseq |