Assay Details
| Gene Symbol: CDYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152342.3 | NP_689555.2 | 4 - 5 | 1108 | 77 | |
| XM_011522866.1 | XP_011521168.1 | 4 - 5 | 1120 | 77 | ||
| XM_011522867.2 | XP_011521169.1 | 4 - 5 | 1663 | 77 | ||
| GenBank mRNA | AK096185.1 | - | 4 - 5 | 1108 | 77 | |
| AX781670.1 | - | 4 - 5 | 1098 | 77 | ||
| AX781671.1 | - | 4 - 5 | 1006 | 77 | ||
| AY273798.1 | - | 4 - 5 | 1148 | 77 | ||
| BC069440.1 | - | 4 - 5 | 1053 | 7262286 | 77 | |
| BC100803.1 | - | 4 - 5 | 1027 | 40000745 | 77 | |
| BC100804.1 | - | 4 - 5 | 1027 | 40000746 | 77 | |
| BC100805.1 | - | 4 - 5 | 1027 | 40000747 | 77 | |
| BC100806.1 | - | 4 - 5 | 1027 | 40000748 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001294.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000567924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000567924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017967.hg.1 | ENST00000299564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017967.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017967.hg.1 | TC16001294.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185233.hg.1 | TC1600010966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000561616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000562812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000563890.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000566173.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000567924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000567924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | ENST00000570137.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010966.hg.1 | NM_152342.3 | Refseq |

CDYL2
Genomic Map