Assay Details
| Gene Symbol: BCL11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018014.3 | NP_060484.2 | 4 - 5 | 2457 | 74 | |
| GenBank mRNA | AB058712.1 | - | 4 - 5 | 2357 | 74 | |
| AF080216.1 | - | 6 - 7 | 2537 | 74 | ||
| AJ404612.1 | - | 4 - 5 | 2457 | 74 | ||
| AK300012.1 | - | 6 - 7 | 2148 | 74 | ||
| BC021098.1 | - | 4 - 5 | 2338 | 5087967 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
protein sumoylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
protein sumoylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000335712.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000356842.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000358510.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000359629.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000409351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000477659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000479026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000479026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000479026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000479026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000489516.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000492272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000492272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000492272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000492272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000492272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | ENST00000631857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_018014.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_022893.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012713.hg.1 | NM_138559.1 | Refseq |

BCL11A
Genomic Map