Assay Details
| Gene Symbol: MINK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170663.4 | NP_733763.1 | 19 - 20 | 2539 | 95 | |
| GenBank mRNA | AB041926.1 | - | 19 - 20 | 2479 | 95 | |
| AB209450.1 | - | 18 - 19 | 3164 | 95 | ||
| AL157418.1 | - | 14 - 15 | 2178 | 95 | ||
More Information
Set Membership: |
|

MINK1
Genomic Map