Assay Details
Gene Symbol: NEK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC112240.1 | - | 1 - 2 | 79 | 8327708 | 115 |
BC113705.1 | - | 1 - 2 | 79 | 8322757 | 115 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC112240.1,BC113705.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000268766.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000543014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000579671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000581000.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000581000.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000581000.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000584342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000592510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000593261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000593261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000593261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000593261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000593261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | ENST00000593261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007385.hg.1 | NM_178170.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 170843 | - | - |
