Assay Details
| Gene Symbol: SMPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318088.1 | NP_001305017.1 | 2 - 3 | 1314 | 144 | |
| GenBank mRNA | HQ132748.1 | - | 2 - 3 | 1117 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SMPD1
Genomic Map