Assay Details
| Gene Symbol: TNRC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012740.1 | XP_016868229.1 | 20 | 6444 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 29 member 4 | SLC29A4 |
ENT4, PMAT |
-23.0kb | Anti-sense | Chr. 7 - 5274311 - 5304073 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100129484 | LOC100129484 | 98.7kb | Sense | Chr. 7 - 5426281 - 5429053 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TNRC18
Genomic Map