Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
TC06001828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000244364.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000312431.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000361203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000370754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000370769.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000370788.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000421834.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
ENST00000446842.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022130.hg.1 |
NM_015548.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000244364.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000312431.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000361203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000370754.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000370788.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000421834.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000482156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000523292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000523597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_001144769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_001144770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_015548.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_183380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000244364.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000312431.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000361203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000370754.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000370788.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000421834.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000482156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000523292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
ENST00000523597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_001144769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_001144770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_015548.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
NM_183380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183508.hg.1 |
TC0600012123.hg.1 |
Clariom D TC |