Assay Details
| Gene Symbol: TTC21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024753.4 | NP_079029.3 | 5 - 6 | 680 | 76 | |
| XM_006712761.1 | XP_006712824.1 | 5 - 6 | 623 | 76 | ||
| XM_011511872.2 | XP_011510174.1 | 5 - 6 | 623 | 76 | ||
| XM_017004967.1 | XP_016860456.1 | 5 - 6 | 623 | 76 | ||
| XM_017004969.1 | XP_016860458.1 | 3 - 4 | 323 | 76 | ||
| GenBank mRNA | AB073395.1 | - | 5 - 6 | 587 | 76 | |
| AK057268.1 | - | 5 - 6 | 618 | 76 | ||
| AK292942.1 | - | 5 - 6 | 623 | 76 | ||
| BC055424.1 | - | 5 - 6 | 680 | 4837489 | 76 | |
| DA798655.1 | - | 3 - 4 | 407 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002495.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017004969.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
regulation of transcription from RNA polymerase II promoter
smoothened signaling pathway
regulation of smoothened signaling pathway
ventricular system development
forebrain dorsal/ventral pattern formation
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium morphogenesis
protein localization to cilium
regulation of transcription from RNA polymerase II promoter
smoothened signaling pathway
regulation of smoothened signaling pathway
ventricular system development
forebrain dorsal/ventral pattern formation
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium morphogenesis
protein localization to cilium
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000497425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000497425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | ENST00000497425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039489.hg.1 | TC02002495.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039489.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039489.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228032.hg.1 | TC0200016747.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000243344.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000464374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000476227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000484129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000497425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000497425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | ENST00000497425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016747.hg.1 | NM_024753.4 | Refseq |

TTC21B
Genomic Map